Human recombinant PDCD10 (from E. coli)
Fournisseur: ProSci Inc.
92-082
PRSI92-082EA 852 EUR InStock
PRSI92-082
Human recombinant PDCD10 (from E. coli)
Protéines et Peptides
Programmed Cell Death Protein 10 (PDCD10) belongs to the PDCD10 family. PDCD10 exists as a homodimer and is widely expressed. PDCD10 can increase mitogen-activated protein kinase activity and MST4 activity. PDCD10 is required for normal cardiovascular development and normal angiogenesis, vasculogenesis and hematopoiesis during embryonic development. Defects in PDCD10 are the cause of cerebral cavernous malformations type 3.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité