Human recombinant STAT3 (from E. coli)
Fournisseur: ProSci Inc.
92-085
PRSI92-085EA 852 EUR InStock
PRSI92-085
Human recombinant STAT3 (from E. coli)
Protéines et Peptides
Signal Transducer and Activator of Transcription 3 (STAT3) belongs to the transcription factor STAT family. STAT3 contains one SH2 domain and is a transcription factor expressed in most cell types. STAT3 is activated by multiple cytokines and growth factors including: IFN-a, IL-10, IL-6, IL-11, IL-12, IL-2, EGF etc. STAT3 functions as signal transducer and transcription activator that mediates cellular responses to interleukins, KITLG/SCF and other growth factors. In addition, STAT3 may also mediate cellular responses to activated FGFR1, FGFR2, FGFR3 and FGFR4.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité