Human recombinant Allograft Inflammatory Factor 1 (from E. coli)
Fournisseur: ProSci Inc.
92-087
PRSI92-087EA 654 EUR InStock
PRSI92-087
Human recombinant Allograft Inflammatory Factor 1 (from E. coli)
Protéines et Peptides
Allograft Inflammatory Factor 1 (AIF1) contains two EF-hand domains and exists as a homodimer. AIF1 can be detected in T-lymphocytes and peripheral blood mononuclear cells. AIF1 functions as actin-binding protein that enhances membrane ruffling and RAC activation and can enhance the actin-bundling activity of LCP1. In addition, AIF1 plays a role in RAC signaling and in phagocytosis and may also in macrophage activation and function. AIF1 promotes the proliferation of vascular smooth muscle cells and of T-lymphocytes and plays a role in vascular inflammation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité