Human recombinant ribulose-phosphate 3-epimerase (from E. coli)
Fournisseur: ProSci Inc.
92-090
PRSI92-090EA 584 EUR InStock
PRSI92-090
Human recombinant ribulose-phosphate 3-epimerase (from E. coli)
Enzymes
Ribulose-Phosphate 3-Epimerase (RPE) is a member of the Ribulose-Phosphate 3-Epimerase family. RPE exists as a homodimer and catalyses the reversible epimerisation of D-ribulose 5-phosphate to D-xylulose 5-phosphate. RPE binds one divalent metal cation per subunit and contains tightly bound Fe2+ when produced in E. coli, but the physiological cofactor may be Co2+, Mn2+ or Zn2+. It has been shown that RPE participates in 3 metabolic pathways: pentose phosphate pathway, pentose and glucuronate interconversions, and carbon fixation.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité