Human recombinant Bisphosphoglycerate Mutase (from E. coli)
Fournisseur: ProSci Inc.
92-092
PRSI92-092EA 584 EUR InStock
PRSI92-092
Human recombinant Bisphosphoglycerate Mutase (from E. coli)
Enzymes
Bisphosphoglycerate Mutase (BPGM) is a member of the Phosphoglycerate Mutase family and BPG-Dependent PGAM subfamily. BPGM is a multifunctional enzyme. BPGM catalyses 2,3-DPG synthesis via its synthetase activity, and 2,3-DPG degradation via its phosphatase activity. It also has phosphoglycerate phosphomutase activity. BPGM plays a major role in regulating hemoglobin oxygen affinity by controlling the levels of 2,3-bisphosphoglycerate (2,3-BPG). Deficiency of BPGM increases the affinity of cells for oxygen and result in hemolytic anemia.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité