Human Recombinant Hepatoma-Derived Growth Factor (from E. coli)
Fournisseur: ProSci Inc.
92-093
PRSI92-093EA 620 EUR InStock
PRSI92-093
Human Recombinant Hepatoma-Derived Growth Factor (from E. coli)
Protéines et Peptides
Hepatoma-Derived Growth Factor is a original member of the HDGF family. HDGF is a cytoplasmic protein and contains one PWWP domain. HDGF expression levels are high in the nucleus and cytoplasm of smooth muscle and endothelial cells. HDGF has proliferative, angiogenic and neurotrophic activity. HDGF was initially characterized as a secreted mitogen from the Huh-7 human hepatoma cell line. As a heparin-binding protein, which is highly expressed in tumor cells where it stimulates proliferation. HDGF has mitogenic activity for fibroblasts and acts as a transcriptional repressor. It has been shown that HDGF is linked with tumorigenesis and the growth of cancer.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité