Human recombinant Lysophospholipase II (from E. coli)
Fournisseur: ProSci Inc.
92-096
PRSI92-096EA 584 EUR InStock
PRSI92-096
Human recombinant Lysophospholipase II (from E. coli)
Enzymes
Acyl-Protein Thioesterase 2 is a cytoplasmic protein that belongs to the AB Hydrolase 2 family. LYPLA2 is a lysophospholipase and hydrolyses fatty acids from S-acylated cysteine residues in proteins such as trimeric G alpha proteins or HRAS. LYPLA2 has hydrolase activity that converts Palmitoyl-protein to palmitate and protein. LYPLA2 regulates the multifunctional lysophospholipids by acting on biological membranes. LYPLA2 participates in Glycerophospholipid metabolism pathway, fatty acid metabolic process and lipid metabolic process.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité