Human recombinant valosin-containing (from E. coli)
Fournisseur: ProSci Inc.
92-101
PRSI92-101EA 884 EUR InStock
PRSI92-101
Human recombinant valosin-containing (from E. coli)
Protéines et Peptides
Valosin-Containing Protein (VCP) is a nuclear protein that belongs to the AAA ATPase family. VCP is a putative ATP-binding protein involved in vesicle transport and fusion, 26S proteasome function, and assembly of peroxisomes. It is necessary for the fragmentation of Golgi stacks during mitosis and their reassembly after mitosis. VCP has been implicated in a number of cellular events that are regulated during mitosis, including homotypic membrane fusion, spindle pole body function, and ubiquitin-dependent protein degradation. VCP participates in the formation of the transitional endoplasmic reticulum (tER) and regulates E3 ubiquitin-protein ligase activity of RNF19A.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité