Human recombinant ETS1 (from E. coli)
Fournisseur: ProSci Inc.
92-104
PRSI92-104EA 852 EUR InStock
PRSI92-104
Human recombinant ETS1 (from E. coli)
Protéines et Peptides
ETS1 Protein (ETS1) is a nuclear protein that belongs to the ETS family. Members of this family recognize the core consensus DNA sequence GGAA/T in target genes. Proteins function either as transcriptional activators or repressors of numerous genes. They are involved in stem cell development, cell senescence and death, and tumorigenesis. ETS1 is a transcription factor, containing one ETS DNA-binding domain and one PNT (pointed) domain. it has been shown to interact with TTRAP, UBE2I and Death Associated Protein 6.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité