Human recombinant B Lymphocyte Kinase (from E. coli)
Fournisseur: ProSci Inc.
92-107
PRSI92-107EA 584 EUR InStock
PRSI92-107
Human recombinant B Lymphocyte Kinase (from E. coli)
Enzymes
Tyrosine-Protein Kinase Blk (BLK) contains one protein kinase domain, one SH2 domain and one SH3 domain. BLK is a non-receptor tyrosine kinase, which is involved in B-lymphocyte development, differentiation and signaling. B-cell receptor (BCR) signaling requires a tight regulation of several protein tyrosine kinases and phosphatases, and associated coreceptors. Signaling through BLK plays an important role in transmitting signals through surface immunoglobulines and supports the pro-B to pre-B transition, as well as the signaling for growth arrest and apoptosis downstream of B-cell receptor. Defects in BLK are a cause of maturity-onset diabetes of the young type 11 (MODY11).
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité