Human recombinant UBE2B (from E. coli)
Fournisseur: ProSci Inc.
92-111
PRSI92-111EA 654 EUR InStock
PRSI92-111
Human recombinant UBE2B (from E. coli)
Protéines et Peptides
Ubiquitin-Conjugating Enzyme E2 B (UBE2B) belongs to the ubiquitin-conjugating enzyme family. UBE2B can catalyze the covalent attachment of ubiquitin to other proteins, and is essential for the multi-ubiquitination and degradation of N-end rule substrates. UBE2B is indispensability for postreplication repair of UV-damaged DNA and may be involved in neurite outgrowth.Additionally, UBE2B may have a role in sepsis-induced muscle protein proteolysis and cancer-induced cachexia.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité