Human recombinant CRADD (from E. coli)
Fournisseur: ProSci Inc.
92-117
PRSI92-117EA 852 EUR InStock
PRSI92-117
Human recombinant CRADD (from E. coli)
Protéines et Peptides
Death Domain-Containing Protein CRADD (CRADD) is widely expressed in most tissues, with particularly high expression in the adult heart, testis, liver, skeletal muscle, fetal liver, and kidney. CRADD contains one CARD domain that mediates the interaction with caspase-2, and one death domain involved in the binding of RIP protein. CRADD functions as an apoptotic adaptor molecule specific for caspase-2 and FASL/TNF receptor-interacting protein RIP. CRADD induces cell apoptosis/cell death in numerous tissues. Defects in CRADD will result in mental retardation.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité