Human recombinant Astrocytic Phosphoprotein PEA-15 (from E. coli)
Fournisseur: ProSci Inc.
92-118
PRSI92-118EA 654 EUR InStock
PRSI92-118
Human recombinant Astrocytic Phosphoprotein PEA-15 (from E. coli)
Protéines et Peptides
Astrocyticphosphoprotein PEA-15 (PEA15) is a death effector domain (DED)-containing protein. PEA15 is mainly expressed in the central nervous system, principally in astrocytes. Increased PEA15 levels affect tumourigenesis and cancer progression. PEA15 is overexpressed in breast cancers and gliomas as well as in type 2 diabetes. PEA15 blocks Ras-mediated inhibition of integrin activation and modulates the ERK MAP kinase cascade. PEA15 also inhibits RPS6KA3 activities by holding it in the cytoplasm. In addition, PEA15 inhibits both TNFRSF6 and TNFRSF1A mediated CASP8 activity and apoptosis. At present, PEA15 expression is also a significant prognostic marker in ovarian cancer.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité