Human Recombinant Gamma-Synuclein (from E. coli)
Fournisseur: ProSci Inc.
92-120
PRSI92-120EA 343 EUR InStock
PRSI92-120
Human Recombinant Gamma-Synuclein (from E. coli)
Protéines et Peptides
Gamma-Synuclein (SNCG) is a member of the Synuclein protein family. Gamma-Synuclein is mostly expressed in the peripheral nervous system and retina. Gamma-Synuclein plays a role in neurofilament network integrity and may be involved in modulating axonal architecture during development and in the adult. In addition, it may also function in modulating the keratin network in skin. SNCG expression in breast tumours has been as a marker for tumour progression. SNCG is also believed to be involved in the pathogenesis of neurodegenerative diseases.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité