Human recombinant BNIP3 (from E. coli)
Fournisseur: ProSci Inc.
92-129
PRSI92-129EA 761 EUR InStock
PRSI92-129
Human recombinant BNIP3 (from E. coli)
Protéines et Peptides
BCL2/Adenovirus E1B 19 kDa Protein-Iinteracting Protein 3 (BNIP3) is a single-pass membrane protein. BNIP3 is a member of the NIP3 family. BNIP3 contains a single Bcl-2 homology 3 domain and interacts with the E1B 19 kDa protein. BNIP3 have been associated with pro-apoptotic function. BNIP3 is an apoptosis-inducing protein that can overcome BCL2 suppression. It plays a role in repartitioning calcium between the two major intracellular calcium stores in association with BCL2. BNIP3 involved in mitochondrial quality control via its interaction with SPATA18/MIEAP, response to mitochondrial damage, participates to mitochondrial protein catabolic process.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité