Human recombinant Actin-related 8 (from E. coli)
Fournisseur: ProSci Inc.
92-130
PRSI92-130EA 852 EUR InStock
PRSI92-130
Human recombinant Actin-related 8 (from E. coli)
Protéines et Peptides
Actin-Related Protein 8 (ACTR8) is a member of the Actin family. ACTR8 is the first example in actin family that was found to be associated with mitotic chromosomes. ACTR8 plays a vital role in the functional organisation of mitotic chromosomes. This protein is a proposed core component of the chromatin remodelling INO80 complex that is involved in transcriptional regulation, DNA replication and probable DNA repair, and it is required for the recruitment of INO80 (and probably the INO80 complex) to sites of DNA damage.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité