Human recombinant sulphotransferase 1B1 (from E. coli)
Fournisseur: ProSci Inc.
92-131
PRSI92-131EA 789 EUR InStock
PRSI92-131
Human recombinant sulphotransferase 1B1 (from E. coli)
Enzymes
Sulfotransferase Family Cytosolic 1B Member 1 (SULT1B1) is a cytosolic enzyme that belongs to the Sulfotransferase 1 family. Human SULT1B1 is a 296 amino acid protein that is highly expressed in the liver, peripheral blood leukocytes, colon, small intestine, and spleen. SULT1B1 utilises 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor, and it can catalyse the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité