Human recombinant Serpin B5 (from E. coli)
Fournisseur: ProSci Inc.
92-135
PRSI92-135EA 525 EUR InStock
PRSI92-135
Human recombinant Serpin B5 (from E. coli)
Protéines et Peptides
Serpin B5 is a secretive protein that belongs to the Serpin (Serine Protease Inhibitor) family and Ov-serpin subfamily. Serpin B5 is expressed in the prostate, testis, intestine, tongue, lung, and thymus. Serpin B5 exhibits no serine protease inhibitory activity. Serpin B5 also functions as tumour suppressor an angiogenesis inhibitor. It has been shown that Serpin B5 blocks the growth, invasion, and metastatic properties of mammary tumours. Furthermore, high expression of Serpin B5 is linked to squamous cell carcinoma in non-small-cell lung cancer.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité