Human recombinant sialic acid synthase (from E. coli)
Fournisseur: ProSci Inc.
92-137
PRSI92-137EA 584 EUR InStock
PRSI92-137
Human recombinant sialic acid synthase (from E. coli)
Enzymes
Sialic Acid Synthase (NANS) is an enzyme that contains one AFP-like domain. NANS is ubiquitous and plays a role in the biosynthetic pathways of sialic acids. NANS produces N-acetylneuraminic acid (Neu5Ac) and 2-keto-3-deoxy-D-glycero-D-galacto-nononic acid (KDN). It also can use N-acetylmannosamine 6-phosphate and mannose 6-phosphate as substrates to generate phosphorylated forms of Neu5Ac and KDN, respectively.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité