Human Recombinant OBFC1 (from E. coli)
Fournisseur: ProSci Inc.
92-138
PRSI92-138EA 852 EUR InStock
PRSI92-138
Human Recombinant OBFC1 (from E. coli)
Protéines et Peptides
CST Complex Subunit STN1 (OBFC1) is a 368 amino acid protein that contains one OB DNA-binding domain. It is a member of the STN1 family. OBFC1 is component of the CST complex, a complex that binds to single-stranded DNA and is required to protect telomeres from DNA degradation. The CST complex binds single-stranded DNA with high affinity in a sequence-independent manner, while isolated subunits bind DNA with low affinity by themselves. In addition to telomere protection, the CST complex has probably a more general role in DNA metabolism at non-telomeric sites.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité