Human recombinant UBE2C (from E. coli)
Fournisseur: ProSci Inc.
92-139
PRSI92-139EA 654 EUR InStock
PRSI92-139
Human recombinant UBE2C (from E. coli)
Protéines et Peptides
Ubiquitin-Conjugating Enzyme E2 C (UBE2C) is a 179 amino acid enzyme that belongs to the Ubiquitin-Conjugating Enzyme family. UBE2C is highly expressed in tumour tissues and at low levels in most adult normal tissues. UBE2C is required for the destruction of mitotic cyclins and for cell cycle progression. UBE2C accepts Ubiquitin from the E1 complex and catalyses its covalent attachment to other proteins. It acts as an essential factor of the anaphase promoting complex/cyclosome (APC/C), which has E3 ubiquitin ligase activity, and targets for destruction substrates from the preceding mitosis (Cyclin A, Cyclin B, Securin, Geminin).
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité