Human recombinant PDCD5 (from E. coli)
Fournisseur: ProSci Inc.
92-144
PRSI92-144EA 553 EUR InStock
PRSI92-144
Human recombinant PDCD5 (from E. coli)
Protéines et Peptides
Programmed Cell Death Protein 5 (PDCD5) is a member of the PDCD5 family. PDCD5 is expressed in tumour cells during apoptosis, independent of apoptosis-inducing stimuli. This protein may function in the process of apoptosis. PDCD5 is upregulated during apoptosis where it translocates rapidly from the cytoplasm to the nucleus. PDCD5 may play an important regulator of K (lysine) acetyltransferase 5 (a protein involved in transcription, DNA damage response and cell cycle control) by inhibiting its proteasome-dependent degradation. PDCD5 is an important novel protein that regulates both apoptotic and non-apoptotic programmed cell death.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité