Human recombinant Death Receptor 6 (from cells)
Fournisseur: ProSci Inc.
92-146
PRSI92-146EA 461 EUR InStock
PRSI92-146
Human recombinant Death Receptor 6 (from cells)
Protéines et Peptides
Tumour Necrosis Factor Receptor Superfamily Member 21 (TNFRSF21) is a type I transmembrane receptor that includes four extracellular cysteine-rich motifs and a cytoplasmic death domain. DR6 is highly expressed in heart, brain, placenta, pancreas, lymph node, thymus and prostate. DR6 may activate NF-kappa-B and JNK to promote apoptosis and T-cell differentiation. In addition, DR6 binds with N-APP, which is released by the deprivation of Trophic-factor. It triggers caspase activation and degeneration of both neuronal cell bodies (via caspase-3) and axons (via caspase-6). DR6 is also expressed on the tumour cell lines and can be induced by TNF- alpha.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité