Human recombinant Carbonic Anhydrase 14 (from E. coli)
Fournisseur: ProSci Inc.
92-149
PRSI92-149EA 789 EUR InStock
PRSI92-149
Human recombinant Carbonic Anhydrase 14 (from E. coli)
Enzymes
Carbonic Anhydrase 14 (CA14) belongs to the Alpha-Carbonic Anhydrase family. It is highly expressed in all parts of the central nervous system and lowly expressed in adult liver, heart, small intestine, colon, kidney, urinary bladder, and skeletal muscle. CA14 along with other Carbonic Anhydrases (CAs) participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. CA14 is predicted to be a type I membrane protein and catalyses the reversible hydration of carbon dioxide.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité