Human recombinant CLIC1 (from E. coli)
Fournisseur: ProSci Inc.
92-150
PRSI92-150EA 588 EUR InStock
PRSI92-150
Human recombinant CLIC1 (from E. coli)
Protéines et Peptides
Chloride Intracellular Channel Protein 1 (CLIC1) belongs to the Chloride Channel CLIC family and contains one GST C-terminal domain. CLIC1 can be expressed in various cell types, but it is especially prominent in the heart, placenta, liver, kidney, and pancreas. CLIC1 can insert into membranes and form chloride ion channels. The channel activity depends on the pH. CLIC1 membrane insertion seems to be redox-regulated and may occur only under oxidising conditions. CLIC1 is also involved in the regulation of the cell cycle.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité