Human recombinant UBE2V2 (from E. coli)
Fournisseur: ProSci Inc.
92-156
PRSI92-156EA 654 EUR InStock
PRSI92-156
Human recombinant UBE2V2 (from E. coli)
Protéines et Peptides
Ubiquitin-Conjugating Enzyme E2 Variant 2 (UBE2V2) is an enzyme that belongs to the ubiquitin-conjugating enzyme family. UBE2V2 can be detected in the placenta, colon, liver, and skin. It forms a heterodimer with UBE2N. The UBE2V2/UBE2N heterodimer catalyses the synthesis of non-canonical poly-ubiquitin chains and which leads to protein degradation by the proteasome. UBE2V2 mediates transcriptional activation of target genes. It plays a role in the control of progress through the cell cycle and differentiation. It also plays a role in the error-free DNA repair pathway and contributes to the survival of cells after DNA damage.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité