Human recombinant Peroxiredoxin 3 (from E. coli)
Fournisseur: ProSci Inc.
92-157
PRSI92-157EA 654 EUR InStock
PRSI92-157
Human recombinant Peroxiredoxin 3 (from E. coli)
Protéines et Peptides
Thioredoxin-Dependent Peroxide Reductase Mitochondrial (PRDX3) is an enzyme that belongs to the AhpC/TSA family. Human and mouse PRDX3 genes are highly conserved, and they map to the regions syntenic between mouse and human chromosomes. Human PRDX3 protein has an antioxidant function and is localised in the mitochondrion. PRDX3 is involved in redox regulation of the cell. PRDX3 protects radical-sensitive enzymes from oxidative damage by a radical-generating system. It acts synergistically with MAP3K13 to regulate the activation of NF-kappa-B in the cytosol.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité