Human Recombinant GCDH (from E. coli)
Fournisseur: ProSci Inc.
92-163
PRSI92-163EA 654 EUR InStock
PRSI92-163
Human Recombinant GCDH (from E. coli)
Protéines et Peptides
Glutaryl-CoA Dehydrogenase Mitochondrial (GCDH) is an enzyme that acts upon glutaryl-coenzyme A, creating crotonyl-coenzyme A. It plays a role in the metabolism of lysine, hydroxylysine and tryptophan. It uses electron transfer flavoprotein as its electron acceptor. Isoform Short is inactive Glutaryl-CoA and electron-transfer flavoprotein to (E)-but-2-enoyl-CoA, CO2 and reduced electron-transfer flavoprotein. A defect in this enzyme is associated with neurological condition glutaric acidemia type 1 and cause a progressive form of early-onset generalised dystonia.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité