Human recombinant UBE2Z (from E. coli)
Fournisseur: ProSci Inc.
92-164
PRSI92-164EA 409 EUR InStock
PRSI92-164
Human recombinant UBE2Z (from E. coli)
Protéines et Peptides
Ubiquitin-Conjugating Enzyme E2 Z (ZUBE2Z) is a member of the E2 ubiquitin-conjugating enzyme family. ZUBE2Z is widely expressed in many tissues, with high expression found in the placenta, pancreas, spleen, and testis. It is ubiquitinates proteins that catalyze the covalent attachment of ubiquitin to other proteins. It has shown that ZUBE2Z participate in signalling pathways, and may be involved in apoptosis regulation.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité