Human recombinant STAT5B (from E. coli)
Fournisseur: ProSci Inc.
92-167
PRSI92-167EA 884 EUR InStock
PRSI92-167
Human recombinant STAT5B (from E. coli)
Protéines et Peptides
Signal Transducer and Activator of Transcription 5b (STAT5B) is a member of the STAT family of transcription factors. They are responsible for an array of cellular activities including regulating growth, survival, differentiation, motility, and the immune response. STAT5B mediates the signal transduction triggered by various cell ligands, such as IL2, IL4, CSF1, and different growth hormones. It has been shown to be involved in diverse biological processes, such as TCR signalling, apoptosis, adult mammary gland development, and sexual dimorphism of liver gene expression. Signal transduction and activator of transcription 5 (STAT5) is a member of the Jak/STAT signal transduction pathway and is activated by a variety of cytokines (IL22, IL6). STAT5 has two isoforms (A and B) that share 93% amino acid identity and bind the DNA consensus site TTCN3GAA. STAT5 mediates cytokine signalling by acting as a signal transducer in the cytoplasm and, upon phosphorylation, translocates to the nucleus and activates transcription of specific genes. STAT5 is involved in a wide array of biological processes ranging from regulating apoptosis to adult mammary gland proliferation, differentiation and survival.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité