Human recombinant alpha-Taxilin (from E. coli)
Fournisseur: ProSci Inc.
92-170
PRSI92-170EA 947 EUR InStock
PRSI92-170
Human recombinant alpha-Taxilin (from E. coli)
Protéines et Peptides
alpha-Taxilin belongs to the taxilin family. alpha-Taxilin exists in almost all tissues, with higher expression levels observed in the heart, kidney, liver, and pancreas. alpha-Taxilin binds to the C-terminal coiled coil region of syntaxin family members STX1A, STX3A, and STX4A, but not when these proteins are complexed with SNAP25, VAMP2 or STXBP1, suggesting that it interacts with syntaxins that do not form the SNARE complex. It is shown that alpha-Taxilin plays multiple roles in the generation and maintenance of neurons through modulation of the NAC-mediated translational machinery and/or the syntaxin-mediated vesicle traffic in the soma. In addition, alpha-Taxilin may be involved in intracellular vesicle traffic and potentially in calcium-dependent exocytosis in neuroendocrine cells.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N, C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité