Human recombinant FGF17 (from E. coli)
Fournisseur: ProSci Inc.
92-171
PRSI92-171EA 654 EUR InStock
PRSI92-171
Human recombinant FGF17 (from E. coli)
Protéines et Peptides
Fibroblast Growth Factor 17 (FGF17) is a member of the heparin-binding growth factors family that is prominently expressed in the cerebellum and cortex. Proteins of this family possess broad mitogenic and cell survival activities and they are involved in a variety of biological processes including embryonic development cell growth, morphogenesis, tissue repair, tumour growth, and invasion. FGF17 plays an important role in the regulation of embryonic development and it acts as signalling molecule in the induction and patterning of the embryonic brain. In addition, FGF17 stimulates the proliferation and activation of cells that express FGF receptors.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité