Human recombinant parvulin-14 (from E. coli)
Fournisseur: ProSci Inc.
92-173
PRSI92-173EA 654 EUR InStock
PRSI92-173
Human recombinant parvulin-14 (from E. coli)
Protéines et Peptides
Peptidyl-prolyl cis-trans isomerase NIMA-interacting 4(PIN4) is a peptidyl-prolyl cis/trans isomerase (PPIase) which interacts with NIMA and is vital for cell cycle regulation. PIN4 has 2 different isoforms: PAR14 and PAR17. Furthermore, PIN4 protein binds to double-stranded DNA under physiological salt conditions. PIN4 is involved as a ribosomal RNA processing factor in ribosome biogenesis. The PAR14 binds to tightly bent AT-rich stretches of double-stranded DNA, but PAR17 binds to double-stranded DNA.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité