Human recombinant Syntenin 1 (from E. coli)
Fournisseur: ProSci Inc.
92-175
PRSI92-175EA 584 EUR InStock
PRSI92-175
Human recombinant Syntenin 1 (from E. coli)
Protéines et Peptides
Syntenin-1 is a molecule linking syndecan-mediated signalling to the cytoskeleton. Syntenin-1 is primarily localised to membrane-associated adherens junctions and focal adhesions but also found at the endoplasmic reticulum and nucleus. The syntenin protein contains tandemly repeated PDZ domains that bind the cytoplasmic, C-terminal domains of a variety of transmembrane proteins. Syntenin-1 may affect cytoskeletal-membrane organisation, cell adhesion, protein trafficking, and the activation of transcription factors. It seems to function as an adapter protein, in adherens junctions may function to couple syndecans to cytoskeletal proteins or signalling components. Syntenin-1 seems to couple transcription factor SOX4 to the IL-5 receptor (IL5RA) and play a role in vesicular trafficking.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité