Human recombinant ABCB5 (from E. coli)
Fournisseur: ProSci Inc.
92-176
PRSI92-176EA 852 EUR InStock
PRSI92-176
Human recombinant ABCB5 (from E. coli)
Protéines et Peptides
ATP-binding cassette sub-family B member 5(ABCB5) is a plasma membrane-spanning protein. ABCB5 is principally expressed in physiological skin and human malignant melanoma. ABCB5 has been suggested to regulate skin progenitor cell fusion and mediate chemotherapeutic drug resistance in stem-like tumour cell subpopulations in human malignant melanoma. It is commonly over-expressed on circulating melanoma tumour cells. Furthermore, the ABCB5+ melanoma- initiating cells were demonstrated to express FLT1 (VEGFR1) receptor tyrosine kinase which was functionally required for efficient xenograft tumour formation, as demonstrated by shRNA knockdown experiments.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-Trx tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité