Human Recombinant NEDD8 (from E. coli)
Fournisseur: ProSci Inc.
92-177
PRSI92-177EA 343 EUR InStock
PRSI92-177
Human Recombinant NEDD8 (from E. coli)
Protéines et Peptides
Human NEDD8 shares 60% amino acid sequence identity to ubiquitin. The only known substrates of NEDD8 modification are the cullin subunits of SCF ubiquitin E3 ligases. The NEDDylation of cullins is critical for the recruitment of E2 to the ligase complex, thus facilitating ubiquitin conjugation. NEDD8 modification has therefore been implicated in cell cycle progression and cytoskeletal regulation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His, SUMO tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité