Human recombinant UBE2M (from E. coli)
Fournisseur: ProSci Inc.
92-178
PRSI92-178EA 306 EUR InStock
PRSI92-178
Human recombinant UBE2M (from E. coli)
Protéines et Peptides
UBE2M is a member of the E2 ubiquitin-conjugating enzyme family. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This protein is linked with a ubiquitin-like protein, NEDD8, which can be conjugated to cellular proteins, such as Cdc53/culin. UBE2M accepts the ubiquitin-like protein NEDD8 from the UBA3-NAE1 E1 complex and catalyses its covalent attachment to other proteins. The specific interaction with the E3 ubiquitin ligase RBX1, but not RBX2, suggests that the RBX1-UBE2M complex neddylates specific target proteins, such as CUL1, CUL2, CUL3 and CUL4. It involved in cell proliferation and is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité