Human recombinant ASB13 (from E. coli)
Fournisseur: ProSci Inc.
92-182
PRSI92-182EA 884 EUR InStock
PRSI92-182
Human recombinant ASB13 (from E. coli)
Protéines et Peptides
Ankyrin repeat and SOCS box protein 13(ASB13) is a member of the ankyrin repeat and SOCS box-containing (ASB) family. ASB13 contain six ankyrin repeats sequence and a SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. ASB13 may be a substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité