Human Recombinant EGR1 (from E. coli)
Fournisseur: ProSci Inc.
92-184
PRSI92-184EA 947 EUR InStock
PRSI92-184
Human Recombinant EGR1 (from E. coli)
Protéines et Peptides
EGR-1 belongs to the EGR family of C2H2-type zinc finger proteins. It is a nuclear protein and functions as a transcriptional regulator. EGR-1 recognises and binds to the DNA sequence 5'-CGCCCCCGC-3'(EGR-site).The products of target genes it activates are required for differentiation and mitogenesis. Studies suggest this is a tumour suppressor gene. EGR-1 has a distinct pattern of expression in the brain, and its induction has been shown to be associated with neuronal activity. Several studies suggest it has a role in neuronal plasticity. EGR-1 has also been found to regulate the expression of synaptobrevin II (a protein important for synaptic exocytosis).
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité