Human recombinant Autophagy 5 (from E. coli)
Fournisseur: ProSci Inc.
92-187
PRSI92-187EA 654 EUR InStock
PRSI92-187
Human recombinant Autophagy 5 (from E. coli)
Protéines et Peptides
ATG5 is an E2 ubiquitin ligase which is necessary for autophagy. Its expression is a relatively late event in the apoptotic process, occurring downstream of caspase activity, dramatically highly expressed in apoptotic cells. It is activated by ATG7, conjugates to ATG12 and associates with isolation membrane to form cup-shaped isolation membrane and autophagosome. The conjugate complex detaches from the membrane immediately before or after autophagosome formation is completed. ATG5 plays an important role in the apoptotic process, possibly within the modified cytoskeleton.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité