Human recombinant CLIC3 (from E. coli)
Fournisseur: ProSci Inc.
92-188
PRSI92-188EA 884 EUR InStock
PRSI92-188
Human recombinant CLIC3 (from E. coli)
Protéines et Peptides
Chloride intracellular channel protein 3 (CLIC3) is encoded by the CLIC3 gene. CLIC3 is a single-pass membrane protein which belongs to the chloride channel CLIC family. It contains one GST C-terminal domain and one GST N-terminal domain. Chloride intracellular channel protein 3 high expressed in the placental, lung and heart, low expressed in skeletal muscle, kidney and pancreas. Chloride intracellular channel protein 3 can insert into membranes and forms chloride ion channels, may participate in cellular growth control.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité