Human recombinant Syntaxin 8 (from E. coli)
Fournisseur: ProSci Inc.
92-190
PRSI92-190EA 588 EUR InStock
PRSI92-190
Human recombinant Syntaxin 8 (from E. coli)
Protéines et Peptides
Syntaxin-8 is a single-pass type IV membrane protein which belongs to the syntaxin family. It contains one t-SNARE coil homology domain. STX8 is highly expressed in heart, also found in brain, kidney, liver, lung, placenta, skeletal muscle, spleen and pancreas. STX8 is involved in protein trafficking from early to late endosomes via vesicle fusion and exocytosis. It as a vesicle trafficking protein functions in the early secretory pathway, possibly mediating retrograde transport form cis-golgi membrane to the ER.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité