Human recombinant SF-20 (from E. coli)
Fournisseur: ProSci Inc.
92-191
PRSI92-191EA 343 EUR InStock
PRSI92-191
Human recombinant SF-20 (from E. coli)
Protéines et Peptides
C19orf10 is a secreted protein which belongs to the UPF0556 family. It is expressed in synovial tissue and detected in synovial fluid of patients with arthropaties. C19orf10 plays a role in proliferation of lymphoid cells and is considered an interleukin.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité