Human recombinant Calreticulin 2 (from E. coli)
Fournisseur: ProSci Inc.
92-193
PRSI92-193EA 553 EUR InStock
PRSI92-193
Human recombinant Calreticulin 2 (from E. coli)
Protéines et Peptides
Calreticulin-3 belongs to the calreticulin family, members of which are calcium binding chaperones localised mainly in the endoplasmic reticulum. It can be divided into a N-terminal globular domain, a proline-rich P-domain forming an elongated arm-like structure and a C-terminal acidic domain. During spermatogenesis process, Calreticulin-3 may act as a lectin-independent chaperone for specific client proteins such as ADAM3. Defects in CALR3 are the cause of familial hypertrophic cardiomyopathy type 19 (CMH19), it is a hereditary heart disorder characterised by ventricular hypertrophy, which is usually asymmetric and often involves the interventricular septum. The symptoms include dyspnea, syncope, collapse, palpitations, and chest pain.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité