Human Recombinant GAMT (from E. coli)
Fournisseur: ProSci Inc.
92-194
PRSI92-194EA 654 EUR InStock
PRSI92-194
Human Recombinant GAMT (from E. coli)
Protéines et Peptides
GAMT is a methyltransferase which belongs to the class I-like SAM-binding methyltransferase superfamily. It contains one RMT2 (arginine N-methyltransferase 2-like) domain and is expressed in liver. GAMT converts guanidoacetate to creatine, using S-adenosylmethionine as the methyl donor. Defects in GAMT are the cause of guanidinoacetate methyltransferase deficiency, which is an autosomal recessive disorder characterised by developmental delay/regression, mental retardation, severe disturbance of expressive and cognitive speech, intractable seizures and movement disturbances, severe depletion of creatine/phosphocreatine in the brain, and accumulation of guanidinoacetic acid in brain and body fluids.
Fusion-Tag: N, C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité