Human Recombinant NECAP2 (from E. coli)
Fournisseur: ProSci Inc.
92-195
PRSI92-195EA 584 EUR InStock
PRSI92-195
Human Recombinant NECAP2 (from E. coli)
Protéines et Peptides
NECAP2 belongs to the NECAP family. The WXXF motifs mediate binding of accessory proteins to the ear-domain of AP-1, GGAs and AP-2 through hydrophobic interactions. Adaptin ear-binding coat-associated protein 2 can interacts with AP1G1 and AP2A1 components of the adapter protein complex AP-1 and AP-2. It also interacts with the GAE domain proteins GGA1, GGA2 and GGA3.
Fusion-Tag: N, C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité