Human recombinant FKBP52 (from E. coli)
Fournisseur: ProSci Inc.
92-198
PRSI92-198EA 409 EUR InStock
PRSI92-198
Human recombinant FKBP52 (from E. coli)
Protéines et Peptides
FKBP4 act as a regulator of microtubule dynamics by inhibiting MAPT/TAU ability to promote microtubule assembly. FKBP4 may play a role in the intracellular trafficking of heterooligomeric forms of steroid hormone receptors between cytoplasm and nuclear compartments, it also may have a protective role against oxidative stress in mitochondria. The isomerase activity controls neuronal growth cones via regulation of TRPC1 channel opening.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité