Human recombinant UPRT (from E. coli)
Fournisseur: ProSci Inc.
92-199
PRSI92-199EA 884 EUR InStock
PRSI92-199
Human recombinant UPRT (from E. coli)
Protéines et Peptides
UPRT is a cytoplasmic enzyme which belongs to the UPRTase family. UPRT is highly expressed in leukocytes, liver, spleen and thymus, with lower expression in brain, lung and skeletal muscle. UPRTcatalyses the conversion of uracil and 5-phosphoribosyl-1-R-diphosphate to uridine monophosphate (UMP). This reaction is an important part of nucleotide metabolism, specifically the pyrimidine salvage pathway. UPRT is a potential target for rational design of drugs to treat parasitic infections and cancer
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité