Mouse recombinant IL33 (from E. coli)
Fournisseur: ProSci Inc.
92-200
PRSI92-200EA 584 EUR InStock
PRSI92-200
Mouse recombinant IL33 (from E. coli)
Protéines et Peptides
Mouse Interleukin 33 (IL-33) is a proinflammatory cytokine which may also regulates gene transcription in producer cells. IL-33 is structurally related to IL-1, which induces helper T cells to produce type 2 cytokines and acts through the receptor IL1RL-1. BindingIL-33 to this receptor activates NF-kappa-B and MAP kinases and induces in vitro Th2 cells to produce cytokines. In vivo, IL-33 induces the expression of IL-4, IL-5, IL-13 and also leads to severe pathological changes in mucosal organs.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité