Human recombinant FGF19 (from E. coli)
Fournisseur: ProSci Inc.
92-201
PRSI92-201EA 654 EUR InStock
PRSI92-201
Human recombinant FGF19 (from E. coli)
Protéines et Peptides
Fibroblast growth factor 19 (FGF19) is a secreted protein which belongs to the FGFs family. FGF19 is expressed in fetal brain, cartilage, retina, and adult gall bladder. FGFs modulate cellular activity via at least 5 distinct subfamilies of high-affinity FGF receptors (FGFRs): FGFR-1, -2, -3, and -4, all with intrinsic tyrosine kinase activity. FGFRs can be important for regulation of glucose and lipid homeostasis. FGF19 has important roles as a hormone produced in the ileum in response to bile acid absorption. It has been shown to cause resistance to diet-induced obesity and insulin desensitisation and to improve insulin, glucose, and lipid profiles in diabetic rodents. FGF19 can be considered as a regulator of energy expenditure.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité